Learn More
Accela Chembio Inc 3-hydroxyazetidine Hydrochloride | 25g | 18621-18-6 | MFCD02683887 | 95+% | Shelf Life: 900 Days | Light Sensitive/moisture Sensitive/nitrogen Or Argon/+4
Catalog No. 50353415
Encompass_Preferred
3-hydroxyazetidine Hydrochloride | 25g | 18621-18-6 | MFCD02683887 | 95+% | Shelf Life: 900 Days | Light Sensitive/moisture Sensitive/nitrogen Or Argon/+4
Catalog No. 50-353-415 Supplier Accela Chembio Inc Supplier No. SY00248225G
3-hydroxyazetidine Hydrochloride | 25g | 18621-18-6 | MFCD02683887 | 95+% | Shelf Life: 900 Days | Light Sensitive/moisture Sensitive/nitrogen Or Argon/+4