Learn More
Medchemexpress LLC 3-Hydroxybenzaldehyde | 100-83-4 | 99.9% | C7H6O2 | 10 G
SDP
Catalog No. 5000434661
Encompass_Preferred
3-Hydroxybenzaldehyde (3-HBA) is a precursor compound for various phenolic compounds. It functions as a substrate for aldehyde dehydrogenase (ALDH) in both rats and humans, and is produced by 3-hydroxybenzyl-alcohol dehydrogenase. This compound has demonstrated vasculoprotective effects both in vitro and in vivo, positioning it as a promising subject for atherosclerosis research.
  • Inhibits vascular smooth muscle cells proliferation, cell cycle, and cell migration
  • Inhibits production levels of p-NF-κB and p-p38 increased by TNF-α in human umbilical vein endothelial cells
  • Exhibits anti-thrombic effects in vivo
  • Considered for research on atherosclerosis
Catalog No. 50-004-34661 Supplier Medchemexpress LLC Supplier No. HY7600610G
3-Hydroxybenzaldehyde (3-HBA) is a precursor compound for various phenolic compounds. It functions as a substrate for aldehyde dehydrogenase (ALDH) in both rats and humans, and is produced by 3-hydroxybenzyl-alcohol dehydrogenase. This compound has demonstrated vasculoprotective effects both in vitro and in vivo, positioning it as a promising subject for atherosclerosis research.
  • Inhibits vascular smooth muscle cells proliferation, cell cycle, and cell migration
  • Inhibits production levels of p-NF-κB and p-p38 increased by TNF-α in human umbilical vein endothelial cells
  • Exhibits anti-thrombic effects in vivo
  • Considered for research on atherosclerosis