Learn More
Selleck Chemical LLC 3-Hydroxybenzoic acid
SDP
Catalog No. 5000002981
Encompass_Preferred
3-Hydroxybenzoic acid (m-Hydroxybenzoic acid 3-Carboxyphenol m-Salicylic acid) is used as an intermediate in the synthesis of plasticisers resins pharmaceuticals etc
Catalog No. 50-000-02981 Supplier Selleck Chemical LLC Supplier No. S636725MG
3-Hydroxybenzoic acid (m-Hydroxybenzoic acid 3-Carboxyphenol m-Salicylic acid) is used as an intermediate in the synthesis of plasticisers resins pharmaceuticals etc