Learn More
Medchemexpress LLC 3-Hydroxybenzoic acid | 99-06-9 | 138.12 | 1 G
SDP
Catalog No. 5000445753
Encompass_Preferred
3-Hydroxybenzoic acid (Standard) is the analytical standard of 3-Hydroxybenzoic acid, intended for research and analytical applications.
  • An orally active endogenous metabolite
  • Agonist for GPR81 and GPR109A
  • Acts as stress response desensitizer
  • Can be used for anti-inflammatory and analgesic study
Catalog No. 50-004-45753 Supplier Medchemexpress LLC Supplier No. HYW0040491G
3-Hydroxybenzoic acid (Standard) is the analytical standard of 3-Hydroxybenzoic acid, intended for research and analytical applications.
  • An orally active endogenous metabolite
  • Agonist for GPR81 and GPR109A
  • Acts as stress response desensitizer
  • Can be used for anti-inflammatory and analgesic study