Learn More
Medchemexpress LLC 3-Hydroxybenzoic acid | 99-06-9 | 138.12 | 1 KG
SDP
Catalog No. 5000445670
Encompass_Preferred
3-Hydroxybenzoic acid is an orally active endogenous metabolite that functions as an agonist for GPR81 and GPR109A. It acts as a stress response desensitizer and can be utilized for anti-inflammatory and analgesic studies.
  • Orally active endogenous metabolite
  • Agonist for GPR81 and GPR109A
  • Acts as a stress response desensitizer
  • Used for anti-inflammatory studies
  • Used for analgesic studies
  • Protects against stress-triggered alterations in body weights and core temperatures in animal models
Catalog No. 50-004-45670 Supplier Medchemexpress LLC Supplier No. HYW0040491KG
3-Hydroxybenzoic acid is an orally active endogenous metabolite that functions as an agonist for GPR81 and GPR109A. It acts as a stress response desensitizer and can be utilized for anti-inflammatory and analgesic studies.
  • Orally active endogenous metabolite
  • Agonist for GPR81 and GPR109A
  • Acts as a stress response desensitizer
  • Used for anti-inflammatory studies
  • Used for analgesic studies
  • Protects against stress-triggered alterations in body weights and core temperatures in animal models