Learn More
Medchemexpress LLC 3-Hydroxybenzoic acid | 99-06-9 | 138.12 | 1 ML
SDP
Catalog No. 5000445855
Encompass_Preferred
3-Hydroxybenzoic acid is an orally active endogenous metabolite. It acts as an agonist for GPR81 and GPR109A and functions as a stress response desensitizer. It can be utilized for anti-inflammatory and analgesic studies.
  • Endogenous metabolite
  • Agonist for GPR81 and GPR109A
  • Stress response desensitizer
  • Potential for anti-inflammatory and analgesic studies
  • Orally active
Catalog No. 50-004-45855 Supplier Medchemexpress LLC Supplier No. HYW00404910MM/1ML
3-Hydroxybenzoic acid is an orally active endogenous metabolite. It acts as an agonist for GPR81 and GPR109A and functions as a stress response desensitizer. It can be utilized for anti-inflammatory and analgesic studies.
  • Endogenous metabolite
  • Agonist for GPR81 and GPR109A
  • Stress response desensitizer
  • Potential for anti-inflammatory and analgesic studies
  • Orally active