Learn More
Medchemexpress LLC 3-Hydroxybenzoic acid | 99-06-9 | 138.12 | 10 G
SDP
Catalog No. 5000447963
Encompass_Preferred
3-Hydroxybenzoic acid is an orally active endogenous metabolite that acts as an agonist for GPR81 and GPR109A. It functions as a stress response desensitizer and can be utilized for anti-inflammatory and analgesic studies.
  • Orally active endogenous metabolite
  • Agonist for GPR81 and GPR109A
  • Functions as a stress response desensitizer
  • Utilized for anti-inflammatory and analgesic studies
Catalog No. 50-004-47963 Supplier Medchemexpress LLC Supplier No. HYW00404910G
3-Hydroxybenzoic acid is an orally active endogenous metabolite that acts as an agonist for GPR81 and GPR109A. It functions as a stress response desensitizer and can be utilized for anti-inflammatory and analgesic studies.
  • Orally active endogenous metabolite
  • Agonist for GPR81 and GPR109A
  • Functions as a stress response desensitizer
  • Utilized for anti-inflammatory and analgesic studies