Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Hydroxybenzoic acid | 99-06-9 | 138.12 | 500 G
SDP

Catalog No. 5000449129
Encompass_Preferred

3-Hydroxybenzoic acid is an orally active endogenous metabolite. It acts as an agonist for GPR81 and GPR109A and functions as a stress response desensitizer. It can be utilized for anti-inflammatory and analgesic studies. This product is intended for research use only and not for sale to patients.

  • Appearance: Solid
  • Color: White to off-white
  • Formula: C7H6O3
  • Structure classification: Phenols, monophenols, ketones, aldehydes, acids
  • Initial source: Endogenous metabolite, human gut microbiota metabolites, microorganisms
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 2 years, -20°C for 1 year

Catalog No. 50-004-49129 Supplier Medchemexpress LLC Supplier No. HYW004049500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Hydroxybenzoic acid is an orally active endogenous metabolite. It acts as an agonist for GPR81 and GPR109A and functions as a stress response desensitizer. It can be utilized for anti-inflammatory and analgesic studies. This product is intended for research use only and not for sale to patients.

  • Appearance: Solid
  • Color: White to off-white
  • Formula: C7H6O3
  • Structure classification: Phenols, monophenols, ketones, aldehydes, acids
  • Initial source: Endogenous metabolite, human gut microbiota metabolites, microorganisms
  • Storage (powder): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 2 years, -20°C for 1 year

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.