Learn More
Medchemexpress LLC 3-Hydroxybenzoic acid | 99-06-9 | MFCD00002506 | 138.12 | 50 G
SDP
Catalog No. 5000445858
Encompass_Preferred
3-Hydroxybenzoic acid is an orally active endogenous metabolite that functions as a stress response desensitizer. It acts as an agonist for GPR81 and GPR109A and can be used for anti-inflammatory and analgesic studies.
  • Orally active endogenous metabolite
  • Agonist for GPR81 and GPR109A
  • Functions as a stress response desensitizer
  • Used for anti-inflammatory and analgesic studies
  • Inhibits cAMP accumulation in SK-N-MC cells
  • Inhibits free fatty acid and glycerol release in mature adipocytes
  • Protects against stress-triggered alterations in vivo
Catalog No. 50-004-45858 Supplier Medchemexpress LLC Supplier No. HYW00404950G
Add to Cart
Add to Cart
3-Hydroxybenzoic acid is an orally active endogenous metabolite that functions as a stress response desensitizer. It acts as an agonist for GPR81 and GPR109A and can be used for anti-inflammatory and analgesic studies.
  • Orally active endogenous metabolite
  • Agonist for GPR81 and GPR109A
  • Functions as a stress response desensitizer
  • Used for anti-inflammatory and analgesic studies
  • Inhibits cAMP accumulation in SK-N-MC cells
  • Inhibits free fatty acid and glycerol release in mature adipocytes
  • Protects against stress-triggered alterations in vivo