Learn More
Medchemexpress LLC 3-Hydroxybenzoic acid | 99-06-9 | MFCD00002506 | 138.12 | 500 MG
SDP
Catalog No. 5000447507
Encompass_Preferred
3-Hydroxybenzoic acid is an orally active endogenous metabolite. It acts as an agonist for GPR81 and GPR109A, and functions as a stress response desensitizer. This compound can be utilized for anti-inflammatory and analgesic studies.
  • Acts as an agonist for GPR81 and GPR109A
  • Functions as a stress response desensitizer
  • Used for anti-inflammatory studies
  • Used for analgesic studies
  • Inhibits cAMP accumulation in SK-N-MC cells
  • Inhibits free fatty acid and glycerol release
Catalog No. 50-004-47507 Supplier Medchemexpress LLC Supplier No. HYW004049500MG
3-Hydroxybenzoic acid is an orally active endogenous metabolite. It acts as an agonist for GPR81 and GPR109A, and functions as a stress response desensitizer. This compound can be utilized for anti-inflammatory and analgesic studies.
  • Acts as an agonist for GPR81 and GPR109A
  • Functions as a stress response desensitizer
  • Used for anti-inflammatory studies
  • Used for analgesic studies
  • Inhibits cAMP accumulation in SK-N-MC cells
  • Inhibits free fatty acid and glycerol release