Learn More
Sigma Aldrich Fine Chemicals Biosciences 3 Hydroxybutyrate Dehydrog
Catalog No. 502905335
Encompass
3-Hydroxybutyrate dehydrogenase (3-HBDH) enzyme obtained from Rhodobacter sphaeroides is generally used for the quantification of ketone bodies such as D-3-hydroxybutyrate and acetoacetate.
Catalog No. 50-290-5335 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 10127841001
3-Hydroxybutyrate dehydrogenase (3-HBDH) enzyme obtained from Rhodobacter sphaeroides is generally used for the quantification of ketone bodies such as D-3-hydroxybutyrate and acetoacetate.