Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Hydroxybutyric acid

Catalog No. 1668985G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-689-85G 5 g
16-689-81G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16-689-85G Supplier Sigma Aldrich Supplier No. 1668985G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-hydroxybutyric acid forms films of random copolymer with (r)-3-hydroxypentanoic acid, (r)-3-hydroxyhexanoic acid, 4-hydroxybutyric acid, 6-hydroxyhexanoic acid and (s,s)-lactide by the melt-crystallized method
  • It is an important intermediate for the fine chemical industry.3-hydroxybutyric acid was used in enantioselective synthesis of d-(-)-3-hydroxybutyric acid by escherichia coli.tandem mass spectrometry data independently generated by scripps center for metabolomics is available to view or download in pdf
  • 166898.pdf tested metabolites are featured on scripps center for metabolomics metlin metabolite database
  • To learn more, visit sigma.com/metlin

Specifications

Density 1.126 g/mL (at 25°C (literature))
Boiling Point 118°C to 120°C (2 mmHg)
Molecular Formula C4H8O3
Refractive Index n20/D 1.443 (literature)
Linear Formula CH3CH(OH)CH2COOH
MDL Number MFCD00004546
Quantity 5 g
Synonym (+/-)-3-Hydroxybutanoic acid; DL-beta-Hydroxybutyric acid
Molecular Weight (g/mol) 104.1
Percent Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.