Learn More
Medchemexpress LLC 3-Hydroxybutyric acid-13C4 sodium | 287111-43-7 | 98.0% | 130.06 | 1 MG
SDP
Catalog No. 5000367057
Encompass_Preferred
3-Hydroxybutyric acid-13C4 sodium is a 13C labeled form of 3-Hydroxybutyric acid sodium. This metabolite is elevated in type I diabetes and can modulate the properties of membrane lipids. It is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes as tracers for quantitation during drug development
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-67057 Supplier Medchemexpress LLC Supplier No. HYW010452S11MG
3-Hydroxybutyric acid-13C4 sodium is a 13C labeled form of 3-Hydroxybutyric acid sodium. This metabolite is elevated in type I diabetes and can modulate the properties of membrane lipids. It is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes as tracers for quantitation during drug development
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs