Learn More
Description
- 3-hydroxybutyric acid forms films of random copolymer with (r)-3-hydroxypentanoic acid, (r)-3-hydroxyhexanoic acid, 4-hydroxybutyric acid, 6-hydroxyhexanoic acid and (s,s)-lactide by the melt-crystallized method
- It is an important intermediate for the fine chemical industry.3-hydroxybutyric acid was used in enantioselective synthesis of d-(-)-3-hydroxybutyric acid by escherichia coli.tandem mass spectrometry data independently generated by scripps center for metabolomics is available to view or download in pdf
- 166898.pdf tested metabolites are featured on scripps center for metabolomics metlin metabolite database
- To learn more, visit sigma.com/metlin
Specifications
Specifications
| Density | 1.126 g/mL (at 25°C (literature)) |
| Boiling Point | 118°C to 120°C (2 mmHg) |
| Molecular Formula | C4H8O3 |
| Linear Formula | CH3CH(OH)CH2COOH |
| Refractive Index | n20/D 1.443 (literature) |
| MDL Number | MFCD00004546 |
| Quantity | 1 g |
| Synonym | (+/-)-3-Hydroxybutanoic acid; DL-beta-Hydroxybutyric acid |
| Molecular Weight (g/mol) | 104.1 |
| Percent Purity | 95% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.