Learn More
Selleck Chemical LLC 3-Hydroxybutyric acid-25mg
SDP
Catalog No. 502389889
Encompass_Preferred
3-Hydroxybutyric acid (3-hydroxybutanoic acid, β-Hydroxybutyric acid, BHB) is a small molecule metabolite that is elevated in type I diabetes.,
Catalog No. 50-238-9889 Supplier Selleck Chemical LLC Supplier No. S103125MG
3-Hydroxybutyric acid (3-hydroxybutanoic acid, β-Hydroxybutyric acid, BHB) is a small molecule metabolite that is elevated in type I diabetes.,