Learn More
Medchemexpress LLC 3-Hydroxybutyric acid | 300-85-6 | 99.3% | 104.10 | 500 MG
SDP
Catalog No. 5000437036
Encompass_Preferred
3-Hydroxybutyric acid (β-Hydroxybutyric acid) is an endogenous metabolite that is elevated in type I diabetes. It can modulate the properties of membrane lipids.
  • Endogenous metabolite
  • Can interact with lipids and alter phase behavior at clinical concentrations.
  • Diminishes the interfacial viscosity of DPPC monolayers.
  • For research use only.
Catalog No. 50-004-37036 Supplier Medchemexpress LLC Supplier No. HY113378500MG
3-Hydroxybutyric acid (β-Hydroxybutyric acid) is an endogenous metabolite that is elevated in type I diabetes. It can modulate the properties of membrane lipids.
  • Endogenous metabolite
  • Can interact with lipids and alter phase behavior at clinical concentrations.
  • Diminishes the interfacial viscosity of DPPC monolayers.
  • For research use only.