Learn More
Medchemexpress LLC 3-Hydroxybutyric acid | 300-85-6 | 99.8% | 104.10 | 200 MG
SDP
Supplier:  Medchemexpress LLC HY113378200MG
Encompass_Preferred
3-Hydroxybutyric acid (β-Hydroxybutyric acid) is an endogenous metabolite that is elevated in type I diabetes and can modulate the properties of membrane lipids. It interacts with lipid monolayers at concentrations that impair consciousness and diminishes the interfacial viscosity of DPPC Monolayers. It is for research use only.
  • Metabolite elevated in type I diabetes.
  • Can modulate the properties of membrane lipids.
  • Interacts with lipid monolayers at concentrations that impair consciousness.
  • Diminishes the interfacial viscosity of DPPC Monolayers.
  • For research use only.
Catalog No. 50-004-36645