Learn More
Medchemexpress LLC 3-Hydroxybutyric acid-d4 sodium | 1219804-68-8 | 99.7% | 130.11 | 10 MG
SDP
Catalog No. 5000438651
Encompass_Preferred
The product is a deuterium-labeled form of 3-Hydroxybutyric acid. This compound is an isotope-labeled metabolite that is typically elevated in individuals with type I diabetes. It has been observed to influence the characteristics of membrane lipids. This product is intended solely for research purposes and is not for human use.
Features and Benefits:
  • Isotope-labeled compound for research applications.
  • Can be used as a tracer in various studies.
  • Suitable as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Deuteration may impact the pharmacokinetic and metabolic profiles in drug development processes.
Catalog No. 50-004-38651 Supplier Medchemexpress LLC Supplier No. HY113378S10MG
The product is a deuterium-labeled form of 3-Hydroxybutyric acid. This compound is an isotope-labeled metabolite that is typically elevated in individuals with type I diabetes. It has been observed to influence the characteristics of membrane lipids. This product is intended solely for research purposes and is not for human use.
Features and Benefits:
  • Isotope-labeled compound for research applications.
  • Can be used as a tracer in various studies.
  • Suitable as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Deuteration may impact the pharmacokinetic and metabolic profiles in drug development processes.