Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-hydroxybutyric acid-d4 sodium | 1219804-68-8 | MFCD06658713 | 99.7% | 130.11 g/mol | C4H3D4NaO3 | 5 MG
SDP

Catalog No. 5000264801
Encompass_Preferred

3-Hydroxybutyric acid-d4 sodium is the deuterated sodium salt of β-hydroxybutyric acid supplied as a solid isotopically labeled standard for research use. It is used as an internal standard or tracer in mass spectrometry, metabolic tracing, and quantitative assays to improve measurement accuracy and enable pathway tracking.

  • High isotopic and chemical purity (≈99.7%).
  • Suitable as internal standard for mass spectrometry and quantitative assays.
  • Solid form for convenient handling and storage.
  • Available in milligram-scale quantities for analytical workflows.
  • Molecular weight 130.11 g/mol; formula C4H3D4NaO3.

Catalog No. 50-002-64801 Supplier Medchemexpress LLC Supplier No. HY113378S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Hydroxybutyric acid-d4 sodium is the deuterated sodium salt of β-hydroxybutyric acid supplied as a solid isotopically labeled standard for research use. It is used as an internal standard or tracer in mass spectrometry, metabolic tracing, and quantitative assays to improve measurement accuracy and enable pathway tracking.

  • High isotopic and chemical purity (≈99.7%).
  • Suitable as internal standard for mass spectrometry and quantitative assays.
  • Solid form for convenient handling and storage.
  • Available in milligram-scale quantities for analytical workflows.
  • Molecular weight 130.11 g/mol; formula C4H3D4NaO3.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.