Learn More
Medchemexpress LLC HY-W010452 500mg Medchemexpress, 3-Hydroxybutyric acid (sodium) CAS:150-83-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW010452500MG
Encompass_Preferred
Medchemexpress, HY-W010452 500mg 3-Hydroxybutyric acid (sodium) CAS:150-83-4 3-Hydroxybutyric acid sodium (β-Hydroxybutyric acid sodium) is a metabolite that is elevated in type I diabetes. 3-Hydroxybutyric acid sodium can modulate the properties of membrane lipids. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3295