Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Hydroxybutyric acid sodium | 150-83-4 | 98.0% | 126.09 | 10MM/1ML
SDP

Catalog No. 5000366450
Encompass_Preferred

3-Hydroxybutyric acid sodium is a metabolite that is elevated in type I diabetes. It can modulate the properties of membrane lipids. It is also capable of interacting with lipids (modeled using a DPPC monolayer) and altering phase behavior at clinical concentrations, and diminishes the interfacial viscosity of DPPC Monolayers.

  • Target: Endogenous Metabolite
  • Research Areas: Metabolic Disease
  • Classification of Application Fields: Ketones, Aldehydes, Acids
  • Modulates membrane lipid properties
  • Interacts with lipids and alters phase behavior
  • Diminishes interfacial viscosity of DPPC Monolayers

Catalog No. 50-003-66450 Supplier Medchemexpress LLC Supplier No. HYW01045210MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Hydroxybutyric acid sodium is a metabolite that is elevated in type I diabetes. It can modulate the properties of membrane lipids. It is also capable of interacting with lipids (modeled using a DPPC monolayer) and altering phase behavior at clinical concentrations, and diminishes the interfacial viscosity of DPPC Monolayers.

  • Target: Endogenous Metabolite
  • Research Areas: Metabolic Disease
  • Classification of Application Fields: Ketones, Aldehydes, Acids
  • Modulates membrane lipid properties
  • Interacts with lipids and alters phase behavior
  • Diminishes interfacial viscosity of DPPC Monolayers

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.