Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-hydroxycapric acid | 14292-26-3 | MFCD00133275 | 98.0% | 188.26 g·mol⁻¹ | C10H20O3 | 5 MG
SDP

Catalog No. 5000264336
Encompass_Preferred

3-Hydroxycapric acid (CAS 14292-26-3) is an endogenous medium-chain hydroxy fatty acid used in research as an inhibitor of mitotic progression. Supplied as a white to off-white solid with high purity, it is suitable for in vitro studies in DMSO and can be formulated for in vivo administration. Store the powder at low temperatures to maintain stability.

  • Endogenous medium-chain hydroxy fatty acid.
  • Inhibits mitotic progression in cellular assays.
  • Purity 98.0% as supplied.
  • Molecular weight 188.26 g·mol⁻¹; formula C10H20O3.
  • Soluble in DMSO (100 mg/mL); formulatable for in vivo use.
  • Store powder at -20°C for long-term stability.

Catalog No. 50-002-64336 Supplier Medchemexpress LLC Supplier No. HY1130575MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Hydroxycapric acid (CAS 14292-26-3) is an endogenous medium-chain hydroxy fatty acid used in research as an inhibitor of mitotic progression. Supplied as a white to off-white solid with high purity, it is suitable for in vitro studies in DMSO and can be formulated for in vivo administration. Store the powder at low temperatures to maintain stability.

  • Endogenous medium-chain hydroxy fatty acid.
  • Inhibits mitotic progression in cellular assays.
  • Purity 98.0% as supplied.
  • Molecular weight 188.26 g·mol⁻¹; formula C10H20O3.
  • Soluble in DMSO (100 mg/mL); formulatable for in vivo use.
  • Store powder at -20°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.