Learn More
Cayman Chemical 3-Hydroxycoumarin, Purity: ≥95%, CAS: 939-19-5, 1 g
SDP
Catalog No. 502911063
Encompass_Preferred
A coumarin with diverse biological activities; inhibits 15-LO-1 (IC50 = 9.5 µM); scavenges DPPH radicals in a cell-free assay (IC50 = 0.61 mM); inhibits lipid peroxidation in vitro (IC50 = 69.07 mM); protective against UVB-induced embryotoxicity in sea urchins
Catalog No. 50-291-1063 Supplier Cayman Chemical Supplier No. 348151 G
A coumarin with diverse biological activities; inhibits 15-LO-1 (IC50 = 9.5 µM); scavenges DPPH radicals in a cell-free assay (IC50 = 0.61 mM); inhibits lipid peroxidation in vitro (IC50 = 69.07 mM); protective against UVB-induced embryotoxicity in sea urchins