Learn More
Cayman Chemical 3-HydroxycoumarIn 5g
SDP
Catalog No. 502912675
Encompass_Preferred
A coumarin with diverse biological activities; inhibits 15-LO-1 (IC50 = 9.5 µM); scavenges DPPH radicals in a cell-free assay (IC50 = 0.61 mM); inhibits lipid peroxidation in vitro (IC50 = 69.07 mM); protective against UVB-induced embryotoxicity in sea urchins
Catalog No. 50-291-2675 Supplier Cayman Chemical Supplier No. 348155 G
A coumarin with diverse biological activities; inhibits 15-LO-1 (IC50 = 9.5 µM); scavenges DPPH radicals in a cell-free assay (IC50 = 0.61 mM); inhibits lipid peroxidation in vitro (IC50 = 69.07 mM); protective against UVB-induced embryotoxicity in sea urchins