Learn More
Medchemexpress LLC 3-Hydroxycoumarin | 939-19-5 | 99.3% | 50 MG
SDP
Catalog No. 5000342898
Encompass_Preferred
3-Hydroxycoumarin is an inhibitor of tyrosinase and lipoxygenase (LOX), with an IC50 for recombinant human tyrosinase (rHT) of 26 μM and an IC50 for LOX of 9.5 μM. It exhibits DPPH free radical scavenging activity and has photoprotective effects on sea urchin zygotes and embryos. The compound has also demonstrated anticancer activity against human SK-MEL-28 cells and SK-MEL3 cells in vitro.
  • Inhibits tyrosinase and lipoxygenase
  • Exhibits DPPH free radical scavenging activity
  • Provides photoprotective effects on sea urchin zygotes and embryos
  • Shows anticancer activity against human SK-MEL-28 and SK-MEL3 cells
  • Can be formulated into a vesicle delivery system for topical administration
Catalog No. 50-003-42898 Supplier Medchemexpress LLC Supplier No. HY12717050MG
3-Hydroxycoumarin is an inhibitor of tyrosinase and lipoxygenase (LOX), with an IC50 for recombinant human tyrosinase (rHT) of 26 μM and an IC50 for LOX of 9.5 μM. It exhibits DPPH free radical scavenging activity and has photoprotective effects on sea urchin zygotes and embryos. The compound has also demonstrated anticancer activity against human SK-MEL-28 cells and SK-MEL3 cells in vitro.
  • Inhibits tyrosinase and lipoxygenase
  • Exhibits DPPH free radical scavenging activity
  • Provides photoprotective effects on sea urchin zygotes and embryos
  • Shows anticancer activity against human SK-MEL-28 and SK-MEL3 cells
  • Can be formulated into a vesicle delivery system for topical administration