Learn More
Medchemexpress LLC 3-Hydroxydodecanoic acid | 1883-13-2 | 98.0% | 216.32 | 10 MG
SDP
Catalog No. 5000437260
Encompass_Preferred
3-Hydroxydodecanoic acid is a medium-chain fatty acid. It activates GPR84 and promotes IL-12 production. This compound also exhibits anticancer activity against colon cancer, lung cancer, and melanoma.
  • Medium-chain fatty acid.
  • Activates GPR84.
  • Promotes IL-12 production.
  • Has anticancer activity against colon cancer, lung cancer, and melanoma.
Catalog No. 50-004-37260 Supplier Medchemexpress LLC Supplier No. HY11310710MG
3-Hydroxydodecanoic acid is a medium-chain fatty acid. It activates GPR84 and promotes IL-12 production. This compound also exhibits anticancer activity against colon cancer, lung cancer, and melanoma.
  • Medium-chain fatty acid.
  • Activates GPR84.
  • Promotes IL-12 production.
  • Has anticancer activity against colon cancer, lung cancer, and melanoma.