Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-hydroxydodecanoic acid | 1883-13-2 | MFCD00133279 | 98.0% | 216.32 g·mol⁻1 | C12H24O3 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1131075MG

Encompass_Preferred

3-Hydroxydodecanoic acid is a medium-chain hydroxy fatty acid used as a research reagent and analytical standard. It is reported to activate GPR84 and promote IL-12 production, and has shown anticancer activity in some studies. The compound is supplied as a solid and as a 10 mM solution in DMSO, with a typical stated purity of 98.0%.

  • Purity 98.0%.
  • Molecular formula C12H24O3.
  • Molecular weight 216.32 g·mol⁻1.
  • Available as solid and 10 mM solution in DMSO.
  • Suitable for biological activity studies and analytical standard applications.
  • Soluble in DMSO for assay preparation.

Catalog No. 50-002-64292


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.