Learn More
Selleck Chemical LLC 3-Hydroxyflavone S4790-25mg
SDP
Catalog No. 503111484
Encompass_Preferred
3-Hydroxyflavone (Flavon-3-ol 3-HF 3-Hydroxy-2-phenylchromone) is the simplest molecule of the flavonol class that is usually used in synthesis of biologically active molecules
Catalog No. 50-311-1484 Supplier Selleck Chemical LLC Supplier No. S479025MG
3-Hydroxyflavone (Flavon-3-ol 3-HF 3-Hydroxy-2-phenylchromone) is the simplest molecule of the flavonol class that is usually used in synthesis of biologically active molecules