Learn More
Cayman Chemical 3'-Hydroxygenkwanin, Purity: ≥98%, CAS: 20243-59-8, 50 mg
SDP
Catalog No. 502912581
Encompass_Preferred
A flavonoid with antioxidant and anticancer activities; scavenges superoxide anions in a cell-free assay at 100 µM; decreases EGFR and phosphorylated EGFR levels in, and reduces viability of, PC-9 and H1975 NSCLC cells at 25 and 50 µM; decreases intratumoral levels of EGFR and phosphorylated EGFR and reduces tumor growth in an H1975 mouse xenograft model at 1 mg/kg
Catalog No. 50-291-2581 Supplier Cayman Chemical Supplier No. 3615050 MG
A flavonoid with antioxidant and anticancer activities; scavenges superoxide anions in a cell-free assay at 100 µM; decreases EGFR and phosphorylated EGFR levels in, and reduces viability of, PC-9 and H1975 NSCLC cells at 25 and 50 µM; decreases intratumoral levels of EGFR and phosphorylated EGFR and reduces tumor growth in an H1975 mouse xenograft model at 1 mg/kg