Learn More
Cayman Chemical 3-Hydroxyglutaric Acid, Purity: ≥95%, CAS: 638-18-6, 10 mg
SDP
Catalog No. 502970232
Encompass_Preferred
A metabolite produced during GCDH deficiency and used as a biomarker of glutaric acidemia type 1
Catalog No. 50-297-0232 Supplier Cayman Chemical Supplier No. 1633410 MG
A metabolite produced during GCDH deficiency and used as a biomarker of glutaric acidemia type 1