Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Hydroxyglutaric acid | 638-18-6 | MFCD06208235 | 148.11 | 1G
SDP

Catalog No. 5000447269
Encompass_Preferred

3-Hydroxyglutaric acid is a glutaric acid derivative and an endogenous metabolite. This compound is a white to off-white solid, with a purity of 99.9%. It is classified under ketones, aldehydes, and acids, and is utilized in various disease research fields.

  • High purity of 99.9%
  • Solid, white to off-white appearance
  • Powder storage: -20°C for 3 years; 4°C for 2 years
  • In solvent storage: -80°C for 6 months; -20°C for 1 month
  • Soluble in water at 250 mg/mL and PBS at 100 mg/mL
  • Molecular weight of 148.11

Catalog No. 50-004-47269 Supplier Medchemexpress LLC Supplier No. HYW0082941G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Hydroxyglutaric acid is a glutaric acid derivative and an endogenous metabolite. This compound is a white to off-white solid, with a purity of 99.9%. It is classified under ketones, aldehydes, and acids, and is utilized in various disease research fields.

  • High purity of 99.9%
  • Solid, white to off-white appearance
  • Powder storage: -20°C for 3 years; 4°C for 2 years
  • In solvent storage: -80°C for 6 months; -20°C for 1 month
  • Soluble in water at 250 mg/mL and PBS at 100 mg/mL
  • Molecular weight of 148.11

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.