Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxyglutaric acid, ≥95% (GC), MilliporeSigma™ Supelco™

Catalog No. 111015014 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5014 50 mg
11-101-5013 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-5014 Supplier MilliporeSigma Supelco Supplier No. 0472550MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 638-18-6
Linear Formula C5H8O5
MDL Number MFCD06208235
Synonym beta-Hydroxyglutaric acid; 3-Hydroxypentanedioic acid
Type Analytical Standard
Quantity 50 mg
Formula Weight 148.11
Percent Purity ≥95% (GC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.