Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-hydroxyglutaric acid-d5 | 1219805-72-7 | MFCD08704920 | 99.7% | 153.14 g/mol | C5H3D5O5 | 5 MG
SDP

Catalog No. 5000264253
Encompass_Preferred

3-hydroxyglutaric acid-d5 is the deuterium-labeled analogue of 3-hydroxyglutaric acid supplied as a high-purity solid for use as an analytical standard and internal standard in quantitative mass spectrometry and metabolite profiling. It is intended for method validation and accurate quantitation in biochemical and metabolic studies.

  • High purity (99.7%).
  • Deuterium-labeled for use as an internal standard in mass spectrometry.
  • Solid white to off-white powder physical form.
  • Chemical formula C5H3D5O5; molecular weight 153.14 g/mol.
  • Recommended storage: powder at -20°C (long term) or 4°C (short term).
  • Available in small pack sizes suitable for analytical applications (1-100 mg).

Catalog No. 50-002-64253 Supplier Medchemexpress LLC Supplier No. HY113411S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-hydroxyglutaric acid-d5 is the deuterium-labeled analogue of 3-hydroxyglutaric acid supplied as a high-purity solid for use as an analytical standard and internal standard in quantitative mass spectrometry and metabolite profiling. It is intended for method validation and accurate quantitation in biochemical and metabolic studies.

  • High purity (99.7%).
  • Deuterium-labeled for use as an internal standard in mass spectrometry.
  • Solid white to off-white powder physical form.
  • Chemical formula C5H3D5O5; molecular weight 153.14 g/mol.
  • Recommended storage: powder at -20°C (long term) or 4°C (short term).
  • Available in small pack sizes suitable for analytical applications (1-100 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.