Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Hydroxyhexanoic acid | 10191-24-9 | 96.0% | 132.16 | 1 G
SDP

Catalog No. 5000436534
Encompass_Preferred

3-Hydroxyhexanoic acid is an ester product intended for research use only.

  • Appearance: Liquid (Density: 1.101±0.06 g/cm3)
  • Color: Colorless to light yellow
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage (Pure form): -20°C for 3 years, 4°C for 2 years
  • Storage (In solvent): -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-36534 Supplier Medchemexpress LLC Supplier No. HY1138361G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-Hydroxyhexanoic acid is an ester product intended for research use only.

  • Appearance: Liquid (Density: 1.101±0.06 g/cm3)
  • Color: Colorless to light yellow
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage (Pure form): -20°C for 3 years, 4°C for 2 years
  • Storage (In solvent): -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.