Learn More
Medchemexpress LLC 3-Hydroxyhexanoic acid | 10191-24-9 | 96.0% | 132.16 | 25 MG
SDP
Catalog No. 5000438710
Encompass_Preferred
3-Hydroxyhexanoic acid is an ester product supplied as a colorless to light yellow liquid with a purity of 96.0%. The molecular formula is C6H12O3, and its molecular weight is 132.16. This product is strictly for research use.
  • High purity (96.0%)
  • Available in various quantities, with estimated next-day arrival for select sizes
  • Detailed solubility information and calculators for in vitro and in vivo applications are provided
  • Comprehensive documentation available, including data sheet, COA, SDS, HNMR, MS, and handling instructions
Catalog No. 50-004-38710 Supplier Medchemexpress LLC Supplier No. 25MGHY113836
3-Hydroxyhexanoic acid is an ester product supplied as a colorless to light yellow liquid with a purity of 96.0%. The molecular formula is C6H12O3, and its molecular weight is 132.16. This product is strictly for research use.
  • High purity (96.0%)
  • Available in various quantities, with estimated next-day arrival for select sizes
  • Detailed solubility information and calculators for in vitro and in vivo applications are provided
  • Comprehensive documentation available, including data sheet, COA, SDS, HNMR, MS, and handling instructions