Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-hydroxyhexanoic acid | 10191-24-9 | MFCD02259045 | 78.4% | 132.16 g/mol | C6H12O3 | 5 MG
SDP

Catalog No. 5000263324
Encompass_Preferred

3-hydroxyhexanoic acid is a small hydroxy fatty acid used as a research reagent and reference standard for studies of fatty acid metabolism and analytical method development. It is the 3-hydroxy homolog of hexanoic (caproic) acid and is provided as a solid suitable for biochemical and analytical applications.

  • Used for metabolite profiling and biochemical research.
  • Suitable as a reference standard for analytical methods.
  • Contains hydroxy and carboxylic functional groups (C6H12O3).
  • Molecular weight 132.16 g/mol.
  • Purity approximately 78.4% by GC (batch dependent).
  • Available in small quantities for analytical use.

Catalog No. 50-002-63324 Supplier Medchemexpress LLC Supplier No. HY1138365MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3-hydroxyhexanoic acid is a small hydroxy fatty acid used as a research reagent and reference standard for studies of fatty acid metabolism and analytical method development. It is the 3-hydroxy homolog of hexanoic (caproic) acid and is provided as a solid suitable for biochemical and analytical applications.

  • Used for metabolite profiling and biochemical research.
  • Suitable as a reference standard for analytical methods.
  • Contains hydroxy and carboxylic functional groups (C6H12O3).
  • Molecular weight 132.16 g/mol.
  • Purity approximately 78.4% by GC (batch dependent).
  • Available in small quantities for analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.