Learn More
Cayman Chemical 3-Hydroxyhippuric Acid, Purity: ≥98%, CAS: 1637-75-8, 100 mg
SDP
Catalog No. 502919336
Encompass_Preferred
A polyphenol metabolite; an active metabolite of (+)-catechin; inhibits kynureninase (Ki = 60 µM)
Catalog No. 50-291-9336 Supplier Cayman Chemical Supplier No. 37240100 MG
A polyphenol metabolite; an active metabolite of (+)-catechin; inhibits kynureninase (Ki = 60 µM)