Learn More
Medchemexpress LLC 3-Hydroxyhippuric acid | 1637-75-8 | 99.5% | 195.17 | 10 MM 1 ML
SDP
Catalog No. 5000435749
Encompass_Preferred
3-Hydroxyhippuric acid is an acyl glycine that acts as an inhibitor of kynureninase (Ki = 60 μM). It is found in higher concentrations in individuals with autism spectrum disorders compared to controls and serves as a marker for Clostridium species, showing a positive correlation with their levels. This compound is also a primary metabolite resulting from catechin and wine polyphenol diets.
  • Inhibits kynureninase (Ki = 60 μM).
  • Acts as a marker for Clostridium species.
  • Functions as a primary metabolite from catechin and wine polyphenol diets.
  • Identified in higher concentrations in individuals with autism spectrum disorders.
  • Has a molecular formula of C9H9NO4.
  • Appears as a white to off-white solid.
  • Classified under phenols, monophenols, ketones, aldehydes, and acids.
  • Originates as an endogenous metabolite.
Catalog No. 50-004-35749 Supplier Medchemexpress LLC Supplier No. HY11308510MM/1ML
3-Hydroxyhippuric acid is an acyl glycine that acts as an inhibitor of kynureninase (Ki = 60 μM). It is found in higher concentrations in individuals with autism spectrum disorders compared to controls and serves as a marker for Clostridium species, showing a positive correlation with their levels. This compound is also a primary metabolite resulting from catechin and wine polyphenol diets.
  • Inhibits kynureninase (Ki = 60 μM).
  • Acts as a marker for Clostridium species.
  • Functions as a primary metabolite from catechin and wine polyphenol diets.
  • Identified in higher concentrations in individuals with autism spectrum disorders.
  • Has a molecular formula of C9H9NO4.
  • Appears as a white to off-white solid.
  • Classified under phenols, monophenols, ketones, aldehydes, and acids.
  • Originates as an endogenous metabolite.