Learn More
Medchemexpress LLC 3-hydroxyhippuric acid | 1637-75-8 | 99.7% | 195.17 g/mol | C9H9NO4 | 5 MG
SDP
Catalog No. 5000264295
Encompass_Preferred
3-hydroxyhippuric acid is an analytical standard of an endogenous metabolite used for research and analytical applications. The compound is a white to off-white powder with formula C9H9NO4 and a molecular weight of 195.17 g/mol; recommended storage is powder at -20°C (3 years) or 4°C (2 years).
  • High purity analytical standard (99.74%).
  • Suitable for metabolic profiling and biomarker studies.
  • Provided as a stable powder with defined storage conditions.
  • Certificate of analysis available for batch traceability.
  • Useful as an analytical reference for quantitation and method validation.
Catalog No. 50-002-64295 Supplier Medchemexpress LLC Supplier No. HY113085R5MG
3-hydroxyhippuric acid is an analytical standard of an endogenous metabolite used for research and analytical applications. The compound is a white to off-white powder with formula C9H9NO4 and a molecular weight of 195.17 g/mol; recommended storage is powder at -20°C (3 years) or 4°C (2 years).
  • High purity analytical standard (99.74%).
  • Suitable for metabolic profiling and biomarker studies.
  • Provided as a stable powder with defined storage conditions.
  • Certificate of analysis available for batch traceability.
  • Useful as an analytical reference for quantitation and method validation.