Learn More
Cayman Chemical 3-Hydroxyhippuric Acid, Purity: ≥98%, CAS: 1637-75-8, 250 mg
SDP
Catalog No. 502919370
Encompass_Preferred
A polyphenol metabolite; an active metabolite of (+)-catechin; inhibits kynureninase (Ki = 60 µM)
Catalog No. 50-291-9370 Supplier Cayman Chemical Supplier No. 37240250 MG
A polyphenol metabolite; an active metabolite of (+)-catechin; inhibits kynureninase (Ki = 60 µM)