Learn More
Cayman Chemical 3-Hydroxyhippuric Acid, Purity: ≥98%, CAS: 1637-75-8, 50 mg
SDP
Catalog No. 502915401
Encompass_Preferred
A polyphenol metabolite; an active metabolite of (+)-catechin; inhibits kynureninase (Ki = 60 µM)
Catalog No. 50-291-5401 Supplier Cayman Chemical Supplier No. 3724050 MG
A polyphenol metabolite; an active metabolite of (+)-catechin; inhibits kynureninase (Ki = 60 µM)