Learn More
Selleck Chemical LLC 3-Hydroxyisovaleric acid
SDP
Catalog No. 5000003353
Encompass_Preferred
3-Hydroxyisovaleric acid is a normal human metabolite excreted in the urine
Catalog No. 50-000-03353 Supplier Selleck Chemical LLC Supplier No. S612425UL
3-Hydroxyisovaleric acid is a normal human metabolite excreted in the urine