Learn More
Medchemexpress LLC 3-Hydroxyisovaleric acid | 625-08-1 | 97.0% | 118.13 | 1 ML
SDP
Catalog No. 5000436798
Encompass_Preferred
3-Hydroxyisovaleric acid is an endogenous metabolite normally excreted in urine. Its urinary excretion serves as an early and sensitive indicator of biotin deficiency.
  • Normal endogenous metabolite.
  • Early and sensitive indicator of biotin deficiency.
  • For research use only.
Catalog No. 50-004-36798 Supplier Medchemexpress LLC Supplier No. HY11340910MM/1ML
3-Hydroxyisovaleric acid is an endogenous metabolite normally excreted in urine. Its urinary excretion serves as an early and sensitive indicator of biotin deficiency.
  • Normal endogenous metabolite.
  • Early and sensitive indicator of biotin deficiency.
  • For research use only.