Learn More
Medchemexpress LLC 3-Hydroxyisovaleric acid | 625-08-1 | ≥97.0% | 118.13 | 5 G
SDP
Catalog No. 5000435806
Encompass_Preferred
3-Hydroxyisovaleric acid is a normal endogenous metabolite that is excreted in the urine. Its urinary excretion serves as an early and sensitive indicator of biotin deficiency. This product is categorized as an endogenous metabolite and a disease marker for various conditions.
  • Normal endogenous metabolite
  • Excreted in the urine
  • Early and sensitive indicator of biotin deficiency
  • Disease marker for cancer, endocrine diseases, nervous system disorders, cardiovascular system disorders, and immune system disorders
Catalog No. 50-004-35806 Supplier Medchemexpress LLC Supplier No. HY1134095G
3-Hydroxyisovaleric acid is a normal endogenous metabolite that is excreted in the urine. Its urinary excretion serves as an early and sensitive indicator of biotin deficiency. This product is categorized as an endogenous metabolite and a disease marker for various conditions.
  • Normal endogenous metabolite
  • Excreted in the urine
  • Early and sensitive indicator of biotin deficiency
  • Disease marker for cancer, endocrine diseases, nervous system disorders, cardiovascular system disorders, and immune system disorders