Learn More
Medchemexpress LLC 3-Hydroxyisovaleric acid | 625-08-1 | 97.4% | 118.13 | 10 MG
SDP
Catalog No. 5000435808
Encompass_Preferred
3-Hydroxyisovaleric acid (Standard) is an analytical standard for research and analytical applications. It is an endogenous metabolite found in urine, and its excretion can indicate biotin deficiency.
  • Used for research and analytical applications.
  • Serves as a reference standard for qualitative, quantitative, and methodological research experiments.
  • Compatible with techniques such as HPLC, GC, and MS.
  • A normal endogenous metabolite excreted in urine.
  • Urinary excretion indicates biotin deficiency.
  • Appearance is a colorless to light yellow liquid.
Catalog No. 50-004-35808 Supplier Medchemexpress LLC Supplier No. HY113409R10MG
3-Hydroxyisovaleric acid (Standard) is an analytical standard for research and analytical applications. It is an endogenous metabolite found in urine, and its excretion can indicate biotin deficiency.
  • Used for research and analytical applications.
  • Serves as a reference standard for qualitative, quantitative, and methodological research experiments.
  • Compatible with techniques such as HPLC, GC, and MS.
  • A normal endogenous metabolite excreted in urine.
  • Urinary excretion indicates biotin deficiency.
  • Appearance is a colorless to light yellow liquid.