Learn More
Medchemexpress LLC 3-Hydroxylidocaine 10mg | 34604-55-2 | 250.34 | C14H22N2O2 | 10 MG
SDP
Catalog No. 5000227703
Encompass_Preferred
3-Hydroxylidocaine is an oxidative metabolite of lidocaine provided as a solid research compound. It is used as a reference standard for metabolic, pharmacokinetic, and analytical studies.
  • Suitable as an LC-MS/MS reference standard for quantitative analysis.
  • Relevant metabolite for in vitro and in vivo lidocaine metabolism studies.
  • Supplied as a solid with high purity (≈98.0%).
  • Molecular weight 250.34; formula C14H22N2O2.
  • Intended for research use only; not for human therapeutic use.
Catalog No. 50-002-27703 Supplier Medchemexpress LLC Supplier No. HY11491410MG
3-Hydroxylidocaine is an oxidative metabolite of lidocaine provided as a solid research compound. It is used as a reference standard for metabolic, pharmacokinetic, and analytical studies.
  • Suitable as an LC-MS/MS reference standard for quantitative analysis.
  • Relevant metabolite for in vitro and in vivo lidocaine metabolism studies.
  • Supplied as a solid with high purity (≈98.0%).
  • Molecular weight 250.34; formula C14H22N2O2.
  • Intended for research use only; not for human therapeutic use.