Learn More
Medchemexpress LLC 3-hydroxymandelic acid | 17119-15-2 | MFCD00042723 | 98.2% | 168.15 g/mol | C8H8O4 | 1 ML
SDP
Catalog No. 5000302525
Encompass_Preferred
3-Hydroxymandelic acid is an endogenous metabolite of phenylephrine supplied as a ready-to-use 10 mM solution in DMSO for research applications. It is provided with high reported purity and characterized molecular data; intended for laboratory research use only.
  • High purity (98.2%) for reliable experimental results.
  • Supplied as a 10 mM solution in DMSO for immediate use.
  • Characterized molecular weight of 168.15 g/mol and formula C8H8O4.
  • Applicable to studies of phenylephrine metabolism and endogenous pathways.
  • Storage recommendations support stability in both solvent and powder forms.
Catalog No. 50-003-02525 Supplier Medchemexpress LLC Supplier No. HYW01532610MM/1ML
3-Hydroxymandelic acid is an endogenous metabolite of phenylephrine supplied as a ready-to-use 10 mM solution in DMSO for research applications. It is provided with high reported purity and characterized molecular data; intended for laboratory research use only.
  • High purity (98.2%) for reliable experimental results.
  • Supplied as a 10 mM solution in DMSO for immediate use.
  • Characterized molecular weight of 168.15 g/mol and formula C8H8O4.
  • Applicable to studies of phenylephrine metabolism and endogenous pathways.
  • Storage recommendations support stability in both solvent and powder forms.