Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxymethyl-1-adamantanol, 96%, Thermo Scientific™

Catalog No. AAH6672103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6672103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6672103 Supplier Thermo Scientific Chemicals Supplier No. H6672103

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38584-37-1
Molecular Formula C11H18O2
Molecular Weight (g/mol) 182.263
MDL Number MFCD03105096
InChI Key FORAJDRXEYKDFJ-UHFFFAOYSA-N
Synonym 3-hydroxymethyl adamantan-1-ol, 3-hydroxymethyl-1-adamantanol, tricyclo 3.3.1.13,7 decane-1-methanol, 3-hydroxy, 3-hydroxymethyl-1-adamantol, 3-hydroxy-1-adamantanemethanol, 3-hydroxyadamantyl methanol, 1-hydroxymethyl-3-adamantanol, 3-hydroxy-1-adamantane methanol, 1-hydroxy-3-hydroxymethyladamantane
PubChem CID 12070475
IUPAC Name 3-(hydroxymethyl)adamantan-1-ol
SMILES C1C2CC3(CC1CC(C2)(C3)O)CO

Specifications

Melting Point 158°C to 162°C
Color Brown to White
Quantity 1 g
Formula Weight 182.26
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 3-Hydroxymethyl-1-adamantanol
Hazard Category H318
Precautionary Statement P280-P305+P351+P338-P310
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.