Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Hydroxymethyl)-4-methoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. H15631G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15631G 1 g
H1563-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H15631G Supplier TCI America Supplier No. H15631G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material 3-(Hydroxymethyl)-4-methoxyphenylboronic Acid (contains varying amounts of Anhydride)
CAS 908142-03-0
Color White-Yellow
Molecular Formula C8H11BO4
Quantity 1 g
MDL Number MFCD14155851
Synonym 3-hydroxymethyl-4-methoxyphenyl boronic acid, 3-hydroxymethyl-4-methoxyphenylboronic acid, ksc005q4b, 3-hydroxymethyl-4-methoxybenzeneboronic acid, 3-hydroxymethyl-4-methoxyphenyl boronicacid, b-3-hydroxymethyl-4-methoxyphenyl-boronic acid, boronic acid, b-3-hydroxymethyl-4-methoxyphenyl
InChI Key DOQUCBSZQOPLGT-UHFFFAOYSA-N
SMILES B(C1=CC(=C(C=C1)OC)CO)(O)O
IUPAC Name [3-(hydroxymethyl)-4-methoxyphenyl]boronic acid
Molecular Weight (g/mol) 181.982
PubChem CID 53407782
Formula Weight 181.98
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.