Learn More
Medchemexpress LLC (3-Hydroxymethyl)phenylboronic acid | 87199-15-3 | 151.96 | 25 G
SDP
Catalog No. 5000449283
Encompass_Preferred
3-(Hydroxymethyl)phenylboronic acid is a biochemical reagent suitable for life science-related research as a biological material or organic compound. It is intended for research use only and not for sale to patients.
  • Reactant for copper-mediated trifluoromethylation
  • Reactant for copper-catalyzed transformations from arylboronic acids in water
  • Reactant for Mitsunobu, Suzuki, and amidation reactions with hydroxyphenylamino bromopyrazinecarboxylate
  • Involved in the synthesis of biologically active molecules, including:
    • Mycobacterium tuberculosis H37Rv chorismate mutase inhibitors via Suzuki coupling reactions
    • HIV protease inhibitors with antiviral activity against drug-resistant viruses
    • Pyrrole derivatives for use as PDE4B inhibitors
Catalog No. 50-004-49283 Supplier Medchemexpress LLC Supplier No. 25GHYW007418
3-(Hydroxymethyl)phenylboronic acid is a biochemical reagent suitable for life science-related research as a biological material or organic compound. It is intended for research use only and not for sale to patients.
  • Reactant for copper-mediated trifluoromethylation
  • Reactant for copper-catalyzed transformations from arylboronic acids in water
  • Reactant for Mitsunobu, Suzuki, and amidation reactions with hydroxyphenylamino bromopyrazinecarboxylate
  • Involved in the synthesis of biologically active molecules, including:
    • Mycobacterium tuberculosis H37Rv chorismate mutase inhibitors via Suzuki coupling reactions
    • HIV protease inhibitors with antiviral activity against drug-resistant viruses
    • Pyrrole derivatives for use as PDE4B inhibitors